Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750385.3 | XP_003750433.1 | 4 - 5 | 5966 | 64 | |
XM_006242130.2 | XP_006242192.1 | 5 - 6 | 1023 | 64 | ||
XM_008765806.1 | XP_008764028.1 | 4 - 5 | 1636 | 64 | ||
XM_235497.8 | XP_235497.6 | 5 - 6 | 697 | 64 | ||
GenBank mRNA | AB588919.1 | - | 5 - 6 | 584 | 64 | |
AB588920.1 | - | 4 - 5 | 439 | 64 | ||
AB588921.1 | - | 5 - 6 | 566 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |