Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024759.1 | NP_001019930.1 | 4 - 5 | 929 | 85 | |
XM_006241397.2 | XP_006241459.1 | 4 - 5 | 994 | 85 | ||
GenBank mRNA | BC097355.1 | - | 4 - 5 | 929 | 7441970 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |