Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107680.1 | NP_001101150.1 | 1 - 2 | 121 | 154 | |
XM_006232477.2 | XP_006232539.1 | 1 - 2 | 129 | 154 | ||
XM_006232479.2 | XP_006232541.1 | 1 - 2 | 129 | 154 | ||
GenBank mRNA | FQ215154.1 | - | 1 - 2 | 124 | 154 | |
FQ215514.1 | - | 1 - 2 | 118 | 154 | ||
FQ216377.1 | - | 1 - 2 | 82 | 154 | ||
FQ216411.1 | - | 1 - 2 | 150 | 154 | ||
FQ223879.1 | - | 1 - 2 | 86 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |