Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107680.1 | NP_001101150.1 | 6 - 7 | 690 | 68 | |
XM_006232477.2 | XP_006232539.1 | 5 - 6 | 653 | 68 | ||
XM_006232478.2 | XP_006232540.1 | 591 | 68 | |||
XM_006232479.2 | XP_006232541.1 | 4 - 5 | 569 | 68 | ||
GenBank mRNA | FQ215154.1 | - | 6 - 7 | 693 | 68 | |
FQ215514.1 | - | 6 - 7 | 687 | 68 | ||
FQ216411.1 | - | 6 - 7 | 719 | 68 | ||
FQ223879.1 | - | 6 - 7 | 655 | 68 | ||
FQ224275.1 | - | 3 - 4 | 348 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |