Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109393.1 | NP_001102863.1 | 4 - 5 | 844 | 67 | |
GenBank mRNA | FQ213131.1 | - | 4 - 5 | 581 | 67 | |
FQ214060.1 | - | 4 - 5 | 552 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |