Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047098.2 | NP_001040563.2 | 5 - 6 | 679 | 94 | |
NM_001164091.1 | NP_001157563.1 | 5 - 6 | 695 | 94 | ||
XM_006237367.1 | XP_006237429.1 | 5 - 6 | 703 | 94 | ||
XM_006237368.2 | XP_006237430.1 | 5 - 6 | 889 | 94 | ||
GenBank mRNA | AB259153.1 | - | 4 - 5 | 407 | 94 | |
FQ215513.1 | - | 5 - 6 | 703 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |