Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012813.2 | NP_036945.2 | 1 - 2 | 532 | 66 | |
XM_008763368.1 | XP_008761590.1 | 1 - 2 | 532 | 66 | ||
GenBank mRNA | CB556962.1 | - | 1 - 2 | 536 | 66 | |
U53883.1 | - | 1 - 2 | 253 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |