Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024984.1 | NP_001020155.1 | 7 - 8 | 629 | 86 | |
GenBank mRNA | BC083597.1 | - | 7 - 8 | 629 | 7132841 | 86 |
FQ213743.1 | - | 8 - 9 | 596 | 86 | ||
FQ227558.1 | - | 7 - 8 | 493 | 86 | ||
FQ229098.1 | - | 9 - 10 | 711 | 86 | ||
FQ230170.1 | - | 9 - 10 | 712 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |