Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109352.1 | NP_001102822.1 | 4 - 5 | 1260 | 133 | |
XM_006237311.2 | XP_006237373.1 | 5 - 6 | 1073 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |