Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109352.1 | NP_001102822.1 | 7 - 8 | 2039 | 64 | |
XM_006237311.2 | XP_006237373.1 | 8 - 9 | 1852 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |