Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134327.1 | NP_599154.1 | 4 - 5 | 555 | 76 | |
GenBank mRNA | AF440759.1 | - | 2 - 3 | 290 | 76 | |
BC088219.1 | - | 4 - 5 | 555 | 7384461 | 76 | |
GU357488.1 | - | 4 - 5 | 491 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |