Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192015.1 | NP_001178944.1 | 6 - 7 | 1330 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Probe spans exons |