Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012843.2 | NP_036975.1 | 106 | 100 | ||
XM_006237515.2 | XP_006237577.1 | 215 | 100 | |||
XM_008763369.1 | XP_008761591.1 | 215 | 100 | |||
GenBank mRNA | BC087031.1 | - | 106 | 7192247 | 100 | |
Z54212.1 | - | 75 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |