Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109353.1 | NP_001102823.1 | 3 - 4 | 421 | 129 | |
XM_006237092.2 | XP_006237154.1 | 5 - 6 | 526 | 129 | ||
GenBank mRNA | GU357483.1 | - | 3 - 4 | 408 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |