Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109353.1 | NP_001102823.1 | 4 - 5 | 577 | 65 | |
XM_006237092.2 | XP_006237154.1 | 6 - 7 | 682 | 65 | ||
GenBank mRNA | GU357483.1 | - | 4 - 5 | 564 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |