Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134711.1 | NP_001128183.1 | 3 - 4 | 524 | 120 | |
NM_001134712.1 | NP_001128184.1 | 3 - 4 | 360 | 120 | ||
GenBank mRNA | BC168169.1 | - | 3 - 4 | 524 | 9024666 | 120 |
EV762285.1 | - | 3 - 4 | 391 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |