Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107036.1 | NP_001100506.1 | 8 - 9 | 1659 | 87 | |
XM_006247095.2 | XP_006247157.1 | 13 - 14 | 2116 | 87 | ||
XM_006247096.2 | XP_006247158.1 | 13 - 14 | 2118 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |