Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080583.1 | NP_542150.1 | 11 - 12 | 1582 | 88 | |
XM_008767935.1 | XP_008766157.1 | 12 - 13 | 2339 | 88 | ||
XM_008767936.1 | XP_008766158.1 | 11 - 12 | 1553 | 88 | ||
XM_008767937.1 | XP_008766159.1 | 10 - 11 | 1468 | 88 | ||
XM_008767938.1 | XP_008766160.1 | 10 - 11 | 1485 | 88 | ||
XM_008767939.1 | XP_008766161.1 | 11 - 12 | 2321 | 88 | ||
GenBank mRNA | BC103481.1 | - | 12 - 13 | 1551 | 7929959 | 88 |
M34176.1 | - | 11 - 12 | 1514 | 88 | ||
M77246.1 | - | 11 - 12 | 1582 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |