Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105828.2 | NP_001099298.2 | 4 - 5 | 308 | 99 | |
GenBank mRNA | BC168730.1 | - | 4 - 5 | 308 | 6887426 | 99 |
FQ217757.1 | - | 4 - 5 | 315 | 99 | ||
FQ224188.1 | - | 4 - 5 | 304 | 99 | ||
FQ228457.1 | - | 4 - 5 | 302 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |