Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007804.1 | NP_001007805.1 | 1 - 2 | 56 | 63 | |
GenBank mRNA | BC086364.1 | - | 1 - 2 | 99 | 7304427 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |