Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108279.1 | NP_001101749.1 | 5 - 6 | 1046 | 99 | |
XM_008767841.1 | XP_008766063.1 | 6 - 7 | 1141 | 99 | ||
XM_008767842.1 | XP_008766064.1 | 6 - 7 | 1431 | 99 | ||
XM_008767843.1 | XP_008766065.1 | 6 - 7 | 1131 | 99 | ||
GenBank mRNA | FQ223808.1 | - | 6 - 7 | 1102 | 99 | |
FQ224714.1 | - | 6 - 7 | 1102 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |