Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145755.2 | NP_001139227.2 | 13 - 14 | 3405 | 62 | |
XM_006246755.2 | XP_006246817.1 | 14 - 15 | 3693 | 62 | ||
GenBank mRNA | FJ665183.1 | - | 13 - 14 | 3396 | 62 | |
HM060628.1 | - | 13 - 14 | 3405 | 62 | ||
HM060629.1 | - | 13 - 14 | 3405 | 62 | ||
HM060630.1 | - | 13 - 14 | 3389 | 62 | ||
HM060631.1 | - | 13 - 14 | 3405 | 62 | ||
HM060632.1 | - | 13 - 14 | 3405 | 62 | ||
HM060633.1 | - | 13 - 14 | 3389 | 62 | ||
HM060634.1 | - | 13 - 14 | 3405 | 62 | ||
HM060635.1 | - | 13 - 14 | 3389 | 62 | ||
HM060636.1 | - | 13 - 14 | 3389 | 62 | ||
HM060637.1 | - | 13 - 14 | 3405 | 62 | ||
HM060638.1 | - | 13 - 14 | 3389 | 62 | ||
HM060639.1 | - | 13 - 14 | 3389 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |