Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024488.1 | NP_077814.1 | 10 - 11 | 1070 | 54 | |
GenBank mRNA | AF177476.1 | - | 10 - 11 | 1070 | 54 | |
BC081793.1 | - | 9 - 10 | 992 | 7190258 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |