Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199081.1 | NP_954512.1 | 2 - 3 | 290 | 115 | |
GenBank mRNA | AY349135.1 | - | 2 - 3 | 290 | 115 | |
BC085347.1 | - | 2 - 3 | 325 | 7305181 | 115 | |
D87991.1 | - | 2 - 3 | 283 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |