Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014228.1 | NP_001014250.1 | 7 - 8 | 959 | 95 | |
GenBank mRNA | BC079020.1 | - | 7 - 8 | 959 | 7113829 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |