Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105835.1 | NP_001099305.1 | 1 - 2 | 298 | 64 | |
XM_006247269.2 | XP_006247331.1 | 1 - 2 | 313 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |