Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100158.2 | NP_001093628.1 | 10 - 11 | 1155 | 68 | |
GenBank mRNA | BC105875.1 | - | 10 - 11 | 1100 | 7130292 | 68 |
FQ234161.1 | - | 9 - 10 | 877 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |