Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100158.2 | NP_001093628.1 | 3 - 4 | 368 | 89 | |
GenBank mRNA | BC105875.1 | - | 3 - 4 | 313 | 7130292 | 89 |
CO559861.1 | - | 3 - 4 | 368 | 89 | ||
FQ234161.1 | - | 2 - 3 | 89 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |