Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143895.1 | NP_001137367.1 | 7 - 8 | 935 | 97 | |
XM_008768113.1 | XP_008766335.1 | 6 - 7 | 1359 | 97 | ||
GenBank mRNA | BC161914.1 | - | 7 - 8 | 902 | 8369613 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |