Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013875.1 | NP_001013897.1 | 5 - 6 | 1084 | 80 | |
XM_006251204.2 | XP_006251266.1 | 6 - 7 | 1181 | 80 | ||
XM_006251205.2 | XP_006251267.1 | 5 - 6 | 971 | 80 | ||
XM_006251206.2 | XP_006251268.1 | 6 - 7 | 1014 | 80 | ||
XM_006251207.2 | XP_006251269.1 | 5 - 6 | 972 | 80 | ||
XM_006251208.2 | XP_006251270.1 | 6 - 7 | 1121 | 80 | ||
XM_006251209.2 | XP_006251271.1 | 5 - 6 | 1164 | 80 | ||
XM_006251210.2 | XP_006251272.1 | 4 - 5 | 960 | 80 | ||
GenBank mRNA | BC081912.1 | - | 5 - 6 | 1084 | 7112018 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |