Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760900.1 | XP_008759122.1 | 3 - 4 | 934 | 63 | |
XM_008760901.1 | XP_008759123.1 | 3 - 4 | 897 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |