Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053859.2 | NP_446311.1 | 3 - 4 | 554 | 147 | |
GenBank mRNA | EU253553.1 | - | 4 - 5 | 519 | 147 | |
FQ211811.1 | - | 567 | 147 | |||
U07609.1 | - | 3 - 4 | 557 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |