Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053859.2 | NP_446311.1 | 8 - 9 | 1140 | 88 | |
GenBank mRNA | EU253553.1 | - | 9 - 10 | 1105 | 88 | |
FQ211811.1 | - | 1153 | 88 | |||
U07609.1 | - | 8 - 9 | 1143 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |