Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008724.1 | NP_001008724.1 | 5 - 6 | 556 | 114 | |
NM_052797.1 | NP_434684.1 | 4 - 5 | 556 | 114 | ||
XM_006232532.2 | XP_006232594.1 | 4 - 5 | 600 | 114 | ||
XM_006232533.2 | XP_006232595.1 | 4 - 5 | 592 | 114 | ||
GenBank mRNA | AY310161.1 | - | 5 - 6 | 556 | 114 | |
BC127465.1 | - | 4 - 5 | 556 | 8375711 | 114 | |
FQ209852.1 | - | 4 - 5 | 574 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |