Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rerg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237582.2 | XP_006237644.1 | 4 - 5 | 583 | 86 | |
XM_578417.6 | XP_578417.4 | 4 - 5 | 588 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |