Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237379.2 | XP_006237441.1 | 5 - 6 | 1571 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |