Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173291.1 | NP_775413.1 | 3 - 4 | 319 | 124 | |
GenBank mRNA | AY649838.1 | - | 3 - 4 | 330 | 124 | |
U56822.1 | - | 3 - 4 | 319 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |