Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085405.1 | NP_001078874.1 | 4 - 5 | 407 | 104 | |
XM_008763334.1 | XP_008761556.1 | 4 - 5 | 465 | 104 | ||
XM_008763335.1 | XP_008761557.1 | 4 - 5 | 465 | 104 | ||
GenBank mRNA | DQ113420.1 | - | 4 - 5 | 104 | ||
DQ157011.1 | - | 4 - 5 | 407 | 104 | ||
DQ157012.1 | - | 4 - 5 | 407 | 104 | ||
EF100680.1 | - | 4 - 5 | 407 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |