Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017335.1 | NP_059031.1 | 7 - 8 | 1190 | 92 | |
GenBank mRNA | FQ229573.1 | - | 714 | 92 | ||
U28927.1 | - | 7 - 8 | 1190 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |