Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030838.1 | NP_110465.1 | 9 - 10 | 1308 | 144 | |
XM_006237642.2 | XP_006237704.1 | 11 - 12 | 1523 | 144 | ||
XM_006237643.2 | XP_006237705.1 | 11 - 12 | 1543 | 144 | ||
XM_006237646.2 | XP_006237708.1 | 9 - 10 | 1324 | 144 | ||
XM_006237648.2 | XP_006237710.1 | 6 - 7 | 1736 | 144 | ||
XM_008763432.1 | XP_008761654.1 | 10 - 11 | 2892 | 144 | ||
GenBank mRNA | AF041105.1 | - | 8 - 9 | 1071 | 144 | |
AF083469.1 | - | 9 - 10 | 1308 | 144 | ||
FQ211722.1 | - | 11 - 12 | 1538 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Inventoried Probe spans exons |