Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008281.1 | NP_001008282.1 | 1 - 2 | 357 | 166 | |
XM_006247272.1 | XP_006247334.1 | 1 - 2 | 572 | 166 | ||
GenBank mRNA | BC085881.1 | - | 1 - 2 | 357 | 7191926 | 166 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |