Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011891.1 | NP_001011891.1 | 3 - 4 | 365 | 116 | |
XM_008767558.1 | XP_008765780.1 | 3 - 4 | 378 | 116 | ||
GenBank mRNA | BC079386.1 | - | 3 - 4 | 365 | 7111553 | 116 |
FQ213066.1 | - | 3 - 4 | 379 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |