Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246751.1 | XP_006246813.1 | 2 - 3 | 415 | 76 | |
XM_006246752.2 | XP_006246814.1 | 2 - 3 | 499 | 76 | ||
XM_006246754.2 | XP_006246816.1 | 2 - 3 | 206 | 76 | ||
XM_008767844.1 | XP_008766066.1 | 1 - 2 | 569 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |