Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133320.1 | NP_579854.1 | 6 - 7 | 750 | 79 | |
XM_006246571.2 | XP_006246633.1 | 6 - 7 | 1159 | 79 | ||
XM_006246572.2 | XP_006246634.1 | 6 - 7 | 1152 | 79 | ||
XM_006246573.2 | XP_006246635.1 | 6 - 7 | 900 | 79 | ||
XM_006246574.2 | XP_006246636.1 | 6 - 7 | 785 | 79 | ||
XM_006246575.2 | XP_006246637.1 | 6 - 7 | 1263 | 79 | ||
XM_006246576.2 | XP_006246638.1 | 6 - 7 | 1153 | 79 | ||
XM_006246578.2 | XP_006246640.1 | 6 - 7 | 899 | 79 | ||
GenBank mRNA | AY008298.1 | - | 6 - 7 | 750 | 79 | |
BC070909.1 | - | 6 - 7 | 840 | 7103650 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |