Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133320.1 | NP_579854.1 | 7 - 8 | 846 | 93 | |
XM_006246571.2 | XP_006246633.1 | 7 - 8 | 1255 | 93 | ||
XM_006246572.2 | XP_006246634.1 | 7 - 8 | 1248 | 93 | ||
XM_006246573.2 | XP_006246635.1 | 7 - 8 | 996 | 93 | ||
XM_006246574.2 | XP_006246636.1 | 7 - 8 | 881 | 93 | ||
XM_006246575.2 | XP_006246637.1 | 7 - 8 | 1359 | 93 | ||
XM_006246576.2 | XP_006246638.1 | 7 - 8 | 1249 | 93 | ||
XM_006246578.2 | XP_006246640.1 | 7 - 8 | 995 | 93 | ||
GenBank mRNA | AY008298.1 | - | 7 - 8 | 846 | 93 | |
BC070909.1 | - | 7 - 8 | 936 | 7103650 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |