Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107074.1 | NP_001100544.1 | 1 - 2 | 119 | 110 | |
GenBank mRNA | BC167030.1 | - | 1 - 2 | 91 | 7378211 | 110 |
FQ226313.1 | - | 1 - 2 | 121 | 110 | ||
FQ226643.1 | - | 1 - 2 | 121 | 110 | ||
FQ227263.1 | - | 1 - 2 | 121 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |