Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271597.1 | NP_001258526.1 | 1 - 2 | 147 | 115 | |
NM_001271598.1 | NP_001258527.1 | 1 - 2 | 147 | 115 | ||
GenBank mRNA | FN805189.1 | - | 1 - 2 | 102 | 115 | |
FQ214628.1 | - | 1 - 2 | 128 | 115 | ||
FQ214964.1 | - | 1 - 2 | 127 | 115 | ||
FQ215164.1 | - | 1 - 2 | 128 | 115 | ||
FQ215595.1 | - | 1 - 2 | 128 | 115 | ||
FQ216481.1 | - | 1 - 2 | 128 | 115 | ||
FQ216493.1 | - | 1 - 2 | 128 | 115 | ||
FQ216936.1 | - | 1 - 2 | 128 | 115 | ||
FQ224852.1 | - | 1 - 2 | 128 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |