Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | S79820.1 | - | 2 - 3 | 450 | 68 |
Gene Symbol: LOC690286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001073964.3 | XP_001073964.2 | 2 - 3 | 915 | 68 | |
XM_006247136.1 | XP_006247198.1 | 2 - 3 | 915 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons |