Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105766.1 | NP_001099236.1 | 2 - 3 | 709 | 84 | |
XM_006245868.2 | XP_006245930.1 | 1 - 2 | 362 | 84 | ||
GenBank mRNA | BC169008.1 | - | 2 - 3 | 615 | 7387434 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |