Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012949.2 | NP_037081.2 | 6 - 7 | 494 | 77 | |
XM_006246599.2 | XP_006246661.1 | 6 - 7 | 565 | 77 | ||
XM_006246600.2 | XP_006246662.1 | 6 - 7 | 545 | 77 | ||
GenBank mRNA | BC083566.1 | - | 6 - 7 | 481 | 7193009 | 77 |
DY321069.1 | - | 6 - 7 | 494 | 77 | ||
FQ214810.1 | - | 5 - 6 | 472 | 77 | ||
FQ214945.1 | - | 6 - 7 | 512 | 77 | ||
FQ214984.1 | - | 6 - 7 | 504 | 77 | ||
FQ215034.1 | - | 2 - 3 | 171 | 77 | ||
FQ215197.1 | - | 2 - 3 | 171 | 77 | ||
FQ215221.1 | - | 5 - 6 | 407 | 77 | ||
FQ215310.1 | - | 2 - 3 | 171 | 77 | ||
FQ215357.1 | - | 6 - 7 | 516 | 77 | ||
FQ215526.1 | - | 6 - 7 | 504 | 77 | ||
FQ215607.1 | - | 2 - 3 | 171 | 77 | ||
FQ215648.1 | - | 6 - 7 | 504 | 77 | ||
FQ215759.1 | - | 6 - 7 | 516 | 77 | ||
FQ215780.1 | - | 2 - 3 | 171 | 77 | ||
FQ215843.1 | - | 5 - 6 | 471 | 77 | ||
FQ215865.1 | - | 6 - 7 | 504 | 77 | ||
FQ216020.1 | - | 2 - 3 | 171 | 77 | ||
FQ216271.1 | - | 2 - 3 | 171 | 77 | ||
FQ216382.1 | - | 6 - 7 | 504 | 77 | ||
FQ216386.1 | - | 5 - 6 | 481 | 77 | ||
FQ216660.1 | - | 5 - 6 | 471 | 77 | ||
FQ216781.1 | - | 2 - 3 | 171 | 77 | ||
FQ217095.1 | - | 6 - 7 | 504 | 77 | ||
FQ223831.1 | - | 2 - 3 | 171 | 77 | ||
FQ224938.1 | - | 6 - 7 | 504 | 77 | ||
Y00979.1 | - | 5 - 6 | 456 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |